Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031113.1 | NP_112375.1 | 1 | 463 |
Gene Symbol: LOC100912032 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751421.3 | XP_003751469.1 | 6 | 527 |
Gene Symbol: LOC681260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760378.1 | XP_008758600.1 | 2 | 446 |
Gene Symbol: Rps27l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772051.1 | XP_008770273.1 | 2 | 445 | ||
XM_574121.5 | XP_574121.1 | 2 | 443 |