Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031115.1 | NP_112377.1 | 8 | 1019 | ||
GenBank mRNA | BC081781.1 | - | 8 | 977 | 7189388 | |
X59132.1 | - | 8 | 1019 |
Set Membership: |
G-protein coupled receptor |