Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031236.1 | NP_112515.1 | 2 | 570 | ||
XM_008759420.1 | XP_008757642.1 | 3 | 653 | |||
GenBank mRNA | AB006137.1 | - | 2 | 570 |