Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031334.1 | NP_112624.1 | 4, 5 | 665 | ||
GenBank mRNA | AB017696.1 | - | 4, 5 | 665 |