Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271121.1 | NP_001258050.1 | 3 | 699 | ||
NM_001271122.1 | NP_001258051.1 | 3 | 699 | |||
NM_001271123.1 | NP_001258052.1 | 3 | 699 | |||
NM_001271124.1 | NP_001258053.1 | 3 | 699 | |||
NM_001271125.1 | NP_001258054.1 | 3 | 699 | |||
NM_001271126.1 | NP_001258055.1 | 3 | 699 | |||
NM_001271127.1 | NP_001258056.1 | 3 | 699 | |||
NM_001271128.1 | NP_001258057.1 | 3 | 699 | |||
NM_001271129.1 | NP_001258058.1 | 3 | 699 | |||
NM_001271130.1 | NP_001258059.1 | 3 | 769 | |||
NM_031588.2 | NP_113776.2 | 3 | 699 | |||
GenBank mRNA | AF194993.1 | - | 3 | 769 | ||
AF194995.1 | - | 3 | 602 | |||
AF194996.1 | - | 3 | 602 | |||
AF194997.1 | - | 3 | 602 | |||
AY973241.1 | - | 1 | 91 | |||
AY973243.1 | - | 1 | 91 | |||
AY973245.1 | - | 3 | 367 | |||
AY995221.1 | - | 3 | 355 | |||
AY995222.1 | - | 3 | 355 | |||
AY995223.1 | - | 3 | 355 | |||
M92430.1 | - | 3 | 399 | |||
U02315.1 | - | 3 | 324 | |||
U02316.1 | - | 3 | 699 | |||
U02317.1 | - | 3 | 699 | |||
U02318.1 | - | 3 | 699 | |||
U02319.1 | - | 3 | 699 | |||
U02320.1 | - | 3 | 699 | |||
U02321.1 | - | 3 | 699 | |||
U02322.1 | - | 3 | 699 | |||
U02323.1 | - | 3 | 699 | |||
U02324.1 | - | 3 | 399 |