Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271118.1 | NP_001258047.1 | 9 | 3426 | ||
NM_001271119.1 | NP_001258048.1 | 8 | 3411 | |||
NM_001271120.1 | NP_001258049.1 | 9 | 3553 | |||
NM_001271121.1 | NP_001258050.1 | 10 | 2855 | |||
NM_001271122.1 | NP_001258051.1 | 12 | 3021 | |||
NM_001271125.1 | NP_001258054.1 | 14 | 3177 | |||
NM_001271126.1 | NP_001258055.1 | 13 | 2981 | |||
NM_001271127.1 | NP_001258056.1 | 13 | 3035 | |||
NM_001271128.1 | NP_001258057.1 | 13 | 3108 | |||
NM_001271129.1 | NP_001258058.1 | 12 | 2966 | |||
NM_001271130.1 | NP_001258059.1 | 13 | 3051 | |||
NM_031588.2 | NP_113776.2 | 12 | 2957 | |||
GenBank mRNA | AF194993.1 | - | 13 | 3046 | ||
U02318.1 | - | 12 | 2965 | |||
U02323.1 | - | 12 | 2973 |