Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031643.4 | NP_113831.1 | 5 | 581 | ||
GenBank mRNA | BC089772.1 | - | 5 | 575 | 7382030 | |
D13341.1 | - | 5 | 564 | |||
D14591.1 | - | 5 | 534 | |||
X62313.1 | - | 5 | 571 | |||
Z16415.1 | - | 5 | 564 |