Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031658.1 | NP_113846.1 | 8 | 851 | ||
XM_006246021.2 | XP_006246083.1 | 9 | 802 | |||
XM_006246022.2 | XP_006246084.1 | 9 | 809 | |||
GenBank mRNA | BC070934.1 | - | 8 | 812 | 7098161 | |
D87351.1 | - | 8 | 851 |