Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031664.1 | NP_113852.1 | 7 | 943 | ||
GenBank mRNA | AY029302.1 | - | 7 | 924 | ||
EU032627.2 | - | 7 | 2025 | |||
U25055.1 | - | 7 | 943 | |||
U66723.1 | - | 7 | 914 |
Gene Symbol: LOC102556085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234890.2 | XP_006234952.1 | 2 | 350 |