Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031672.2 | NP_113860.2 | 3 | 449 | ||
GenBank mRNA | BC085327.1 | - | 3 | 449 | 7098404 | |
D63149.1 | - | 3 | 437 |