Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007238.4 | NP_009169.3 | 4 | 830 | ||
NM_183397.2 | NP_899634.1 | 3 | 631 | |||
GenBank mRNA | AF072864.1 | - | 4 | 752 | ||
AK023879.1 | - | 4 | 748 | |||
AK297018.1 | - | 3 | 620 | |||
AK301532.1 | - | 4 | 830 | |||
AL136648.1 | - | 4 | 801 | |||
BC001147.1 | - | 4 | 731 | 3533572 | ||
BQ278083.1 | - | 3 | 599 | |||
CR533471.1 | - | 4 | 707 |