Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031677.1 | NP_113865.1 | 2 | 277 | ||
XM_006244849.2 | XP_006244911.1 | 3 | 2414 | |||
GenBank mRNA | AB008571.1 | - | 2 | 277 |