Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098538.2 | NP_001092008.2 | 3 | 202 | ||
NM_007244.2 | NP_009175.2 | 3 | 434 | |||
GenBank mRNA | AF530472.1 | - | 3 | 416 | ||
AK292934.1 | - | 3 | 438 | |||
AK292991.1 | - | 3 | 430 | |||
AK304321.1 | - | 3 | 380 | |||
BC058035.1 | - | 3 | 437 | 6558043 | ||
BX537839.1 | - | 3 | 196 | |||
DB229285.1 | - | 3 | 438 | |||
S79048.1 | - | 3 | 411 |
Gene Symbol: PRH1-PRR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037918.2 | 9 | 1515 | |||
GenBank mRNA | AB593030.1 | - | 5 | 792 |