Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031685.2 | NP_113873.1 | 3 | 140 | ||
GenBank mRNA | AF007549.1 | - | 3 | 154 | ||
BC061994.1 | - | 3 | 140 | 5599557 | ||
U91539.1 | - | 3 | 139 |