Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031696.1 | NP_113884.1 | 2 | 3328 | ||
XM_006233220.2 | XP_006233282.1 | 2 | 3247 | |||
XM_006233221.2 | XP_006233283.1 | 2 | 3192 | |||
XM_006233222.2 | XP_006233284.1 | 2 | 3189 | |||
XM_008761471.1 | XP_008759693.1 | 3 | 3806 | |||
GenBank mRNA | AB042407.1 | - | 2 | 3328 |
Set Membership: |
G-protein coupled receptor |