Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031708.1 | NP_113896.1 | 2 | 203 | ||
XM_006235822.1 | XP_006235884.1 | 2 | 324 | |||
GenBank mRNA | AB032742.1 | - | 2 | 203 | ||
BC061773.1 | - | 2 | 219 | 5621873 | ||
FQ218007.1 | - | 2 | 221 |