Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031718.1 | NP_113906.1 | 6 | 408 | ||
GenBank mRNA | FQ229541.1 | - | 6 | 610 | ||
J02999.1 | - | 6 | 408 |