Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031719.1 | NP_113907.1 | 7 | 926 | ||
XM_006229792.2 | XP_006229854.1 | 7 | 992 | |||
GenBank mRNA | BC060555.1 | - | 7 | 969 | 6918578 | |
FQ212666.1 | - | 5 | 611 | |||
FQ232880.1 | - | 5 | 612 | |||
L26450.1 | - | 7 | 926 |