Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031735.1 | NP_113923.1 | 3 | 224 | ||
XM_008765458.1 | XP_008763680.1 | 3 | 402 | |||
XM_008765459.1 | XP_008763681.1 | 3 | 402 | |||
XM_008765460.1 | XP_008763682.1 | 3 | 91 | |||
XM_008765461.1 | XP_008763683.1 | 3 | 402 | |||
XM_008765462.1 | XP_008763684.1 | 3 | 402 | |||
GenBank mRNA | AJ001529.1 | - | 3 | 224 | ||
BC161844.1 | - | 3 | 307 | 9027884 |