Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031738.1 | NP_113926.1 | 8 | 993 | ||
XM_008760174.1 | XP_008758396.1 | 2746 | ||||
GenBank mRNA | AF015305.1 | - | 8 | 993 |
Gene Symbol: LOC100911721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749035.3 | XP_003749083.1 | 8 | 1777 | ||
XM_008760194.1 | XP_008758416.1 | 8 | 2746 |