Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271984.1 | NP_001258913.1 | 4 | 263 | ||
NM_031775.3 | NP_113963.2 | 5 | 418 | |||
XM_008761527.1 | XP_008759749.1 | 3 | 208 | |||
XM_008761528.1 | XP_008759750.1 | 3 | 208 | |||
GenBank mRNA | AF025670.1 | - | 5 | 436 | ||
BC078785.1 | - | 5 | 383 | 7123179 | ||
DY310578.1 | - | 4 | 250 | |||
FM068086.1 | - | 4 | 263 |
Gene Symbol: LOC103689977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233278.2 | XP_006233340.1 | 5 | 443 | ||
XM_006233279.2 | XP_006233341.1 | 4 | 265 | |||
XM_008761476.1 | XP_008759698.1 | 5 | 445 | |||
XM_008761477.1 | XP_008759699.1 | 4 | 268 |