Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031832.1 | NP_114020.1 | 5, 6 | 673 | ||
GenBank mRNA | BC089054.1 | - | 5, 6 | 661 | 7312891 | |
FQ210101.1 | - | 5, 6 | 683 | |||
FQ214799.1 | - | 5, 6 | 955 | |||
FQ218337.1 | - | 5, 6 | 673 | |||
FQ219929.1 | - | 5, 6 | 673 | |||
FQ219987.1 | - | 5, 6 | 673 | |||
FQ220157.1 | - | 5, 6 | 673 | |||
FQ220174.1 | - | 5, 6 | 672 | |||
FQ220215.1 | - | 5, 6 | 675 | |||
FQ220224.1 | - | 5, 6 | 667 | |||
FQ220313.1 | - | 5, 6 | 673 | |||
FQ220316.1 | - | 6, 7 | 903 | |||
FQ220348.1 | - | 5, 6 | 673 | |||
FQ220361.1 | - | 5, 6 | 832 | |||
FQ220369.1 | - | 5, 6 | 673 | |||
FQ220439.1 | - | 5, 6 | 672 | |||
FQ220450.1 | - | 5, 6 | 673 | |||
FQ220455.1 | - | 6, 7 | 900 | |||
FQ220487.1 | - | 6, 7 | 873 | |||
FQ220505.1 | - | 5, 6 | 673 | |||
FQ220541.1 | - | 5, 6 | 673 | |||
FQ220563.1 | - | 5, 6 | 673 | |||
FQ220590.1 | - | 5, 6 | 675 | |||
FQ220652.1 | - | 5, 6 | 673 | |||
FQ220701.1 | - | 5, 6 | 672 | |||
FQ220837.1 | - | 5, 6 | 672 | |||
FQ220917.1 | - | 5, 6 | 673 | |||
FQ220938.1 | - | 5, 6 | 673 | |||
FQ220946.1 | - | 5, 6 | 673 | |||
FQ220952.1 | - | 5, 6 | 673 | |||
FQ221377.1 | - | 5, 6 | 673 | |||
FQ223467.1 | - | 5, 6 | 675 | |||
FQ223525.1 | - | 6, 7 | 465 | |||
FQ223531.1 | - | 5, 6 | 673 | |||
FQ229211.1 | - | 5, 6 | 677 | |||
FQ229249.1 | - | 5, 6 | 673 | |||
FQ229259.1 | - | 5, 6 | 673 | |||
FQ229266.1 | - | 5, 6 | 673 | |||
FQ229277.1 | - | 5, 6 | 670 | |||
FQ229326.1 | - | 5, 6 | 673 | |||
FQ229348.1 | - | 5, 6 | 673 | |||
FQ229362.1 | - | 5, 6 | 673 | |||
FQ229490.1 | - | 5, 6 | 673 | |||
FQ229494.1 | - | 5, 6 | 673 | |||
FQ229511.1 | - | 5, 6 | 673 | |||
FQ229560.1 | - | 5, 6 | 675 | |||
FQ229562.1 | - | 5, 6 | 675 | |||
FQ229564.1 | - | 5, 6 | 675 | |||
FQ229577.1 | - | 5, 6 | 673 | |||
FQ229661.1 | - | 5, 6 | 673 | |||
FQ229821.1 | - | 5, 6 | 673 | |||
FQ229967.1 | - | 5, 6 | 675 | |||
FQ229990.1 | - | 5, 6 | 675 | |||
FQ229991.1 | - | 5, 6 | 675 | |||
FQ229995.1 | - | 5, 6 | 673 | |||
FQ230027.1 | - | 5, 6 | 673 | |||
FQ230050.1 | - | 5, 6 | 676 | |||
FQ230072.1 | - | 5, 6 | 673 | |||
FQ230121.1 | - | 5, 6 | 680 | |||
FQ230149.1 | - | 5, 6 | 675 | |||
J02962.1 | - | 5, 6 | 673 | |||
KF639951.1 | - | 4, 5 | 633 | |||
KF639952.1 | - | 4, 5 | 633 | |||
KF639953.1 | - | 4, 5 | 633 | |||
KF639954.1 | - | 4, 5 | 633 | |||
KF639955.1 | - | 5, 6 | 612 | |||
KF639956.1 | - | 5, 6 | 612 | |||
KF639957.1 | - | 5, 6 | 612 | |||
KF639958.1 | - | 5, 6 | 612 | |||
M13697.1 | - | 2, 3 | 261 |