Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031840.1 | NP_114028.1 | 4 | 639 | ||
GenBank mRNA | BC059125.1 | - | 4 | 567 | 6919326 | |
FQ209697.1 | - | 4 | 532 | |||
FQ210400.1 | - | 4 | 531 | |||
FQ211073.1 | - | 5 | 748 | |||
FQ213475.1 | - | 4 | 563 | |||
FQ218913.1 | - | 4 | 533 | |||
FQ218933.1 | - | 4 | 533 | |||
FQ219092.1 | - | 4 | 533 | |||
FQ225110.1 | - | 4 | 601 | |||
M17300.1 | - | 3 | 509 | |||
M34477.1 | - | 4 | 639 |