Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031840.1 | NP_114028.1 | 5 | 769 | ||
GenBank mRNA | BC059125.1 | - | 5 | 697 | 6919326 | |
FQ209697.1 | - | 5 | 662 | |||
FQ210400.1 | - | 5 | 661 | |||
FQ213475.1 | - | 5 | 693 | |||
FQ218913.1 | - | 5 | 663 | |||
FQ218933.1 | - | 5 | 663 | |||
FQ219092.1 | - | 5 | 663 | |||
FQ225110.1 | - | 5 | 731 | |||
M34477.1 | - | 5 | 769 |