Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271091.1 | NP_001258020.1 | 6, 7 | 575 | ||
NM_001271092.1 | NP_001258021.1 | 6, 7 | 575 | |||
NM_001271093.1 | NP_001258022.1 | 6, 7 | 522 | |||
NM_001271094.1 | NP_001258023.1 | 6, 7 | 522 | |||
NM_001271095.1 | NP_001258024.1 | 4, 5 | 440 | |||
NM_031967.2 | NP_114173.2 | 4, 5 | 440 | |||
XM_006255123.2 | XP_006255185.1 | 6, 7 | 517 | |||
GenBank mRNA | AF045564.1 | - | 4, 5 | 372 | ||
AY217029.1 | - | 4, 5 | 411 | |||
AY217030.1 | - | 6, 7 | 522 | |||
AY217031.1 | - | 6, 7 | 522 | |||
AY217032.1 | - | 6, 7 | 570 | |||
AY217033.1 | - | 6, 7 | 570 | |||
AY255791.1 | - | 4, 5 | 411 | |||
FM054107.1 | - | 4, 5 | 393 |