Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031983.2 | NP_114189.1 | 3, 4 | 522 | ||
GenBank mRNA | AB003504.1 | - | 3, 4 | 252 | ||
BC062063.1 | - | 3, 4 | 522 | 5599204 |