Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031987.1 | NP_114193.1 | 13 | 1383 | ||
XM_006236004.2 | XP_006236066.1 | 13 | 1543 | |||
XM_006236005.2 | XP_006236067.1 | 14 | 2873 | |||
GenBank mRNA | BC074004.1 | - | 13 | 1403 | 7098081 | |
J02844.1 | - | 1824 | ||||
U26033.1 | - | 13 | 1383 |