Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032067.1 | NP_114456.1 | 2 | 729 | ||
GenBank mRNA | BC089788.1 | - | 2 | 783 | 7374343 | |
U28830.1 | - | 2 | 733 | |||
U82623.1 | - | 2 | 729 |
Gene Symbol: LOC501456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771014.1 | XP_008769236.1 | 2 | 793 |
Gene Symbol: LOC680627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771013.1 | XP_008769235.1 | 14 | 2640 |