Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033352.1 | NP_203503.1 | 1 | 471 | ||
GenBank mRNA | AF131294.1 | - | 1 | 471 |