Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052809.1 | NP_434696.1 | 2, 3 | 445 | ||
XM_006254698.2 | XP_006254760.1 | 2, 3 | 670 | |||
GenBank mRNA | FQ218376.1 | - | 1, 2 | 82 | ||
FQ219282.1 | - | 1, 2 | 82 | |||
M35266.1 | - | 2, 3 | 445 |