Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007351.2 | NP_031377.2 | 6 | 2347 | ||
XM_005262856.2 | XP_005262913.1 | 5 | 2267 | |||
XM_017007893.1 | XP_016863382.1 | 7 | 2588 | |||
GenBank mRNA | AK302421.1 | - | 6 | 1726 | ||
AK313566.1 | - | 6 | 2372 | |||
BC063848.1 | - | 6 | 2347 | 6180674 | ||
U27109.1 | - | 6 | 2347 |