Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053495.1 | NP_445947.1 | 2 | 171 | ||
XM_008767042.1 | XP_008765264.1 | 4 | 695 | |||
XM_008767043.1 | XP_008765265.1 | 3 | 581 | |||
XM_008767044.1 | XP_008765266.1 | 3 | 581 | |||
GenBank mRNA | AB002801.1 | - | 2 | 171 | ||
AJ272428.1 | - | 4 | 458 | |||
AJ272429.1 | - | 3 | 344 |