Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053584.2 | NP_446036.1 | 9 | 1551 | ||
XM_006246914.1 | XP_006246976.1 | 9 | 1619 | |||
XM_008768132.1 | XP_008766354.1 | 8 | 1422 | |||
GenBank mRNA | CB789285.1 | - | 1 | 212 |