Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053607.1 | NP_446059.1 | 3 | 323 | ||
XM_008760551.1 | XP_008758773.1 | 3 | 461 | |||
GenBank mRNA | AB012933.1 | - | 3 | 323 | ||
BC072497.1 | - | 3 | 315 | 7096959 | ||
FQ218149.1 | - | 3 | 378 |