Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053617.2 | NP_446069.1 | 3 | 205 | ||
GenBank mRNA | BC091133.1 | - | 3 | 169 | ||
BC107447.1 | - | 3 | 198 | 7323668 | ||
DY312674.1 | - | 3 | 205 | |||
FQ218610.1 | - | 3 | 210 | |||
FQ218635.1 | - | 3 | 159 | |||
FQ218700.1 | - | 3 | 210 | |||
FQ219645.1 | - | 3 | 277 |