Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053619.1 | NP_446071.1 | 2 | 1647 | ||
GenBank mRNA | AB003042.1 | - | 2 | 1647 | ||
BC078770.1 | - | 2 | 1630 | 7121456 | ||
Y09613.1 | - | 2 | 1631 |
Set Membership: |
G-protein coupled receptor |