Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053631.3 | NP_446083.1 | 3 | 535 | ||
XM_006230630.2 | XP_006230692.1 | 3 | 548 | |||
XM_006230631.1 | XP_006230693.1 | 3 | 572 | |||
XM_006230632.2 | XP_006230694.1 | 3 | 545 | |||
GenBank mRNA | AB024333.1 | - | 3 | 530 | ||
BC084726.1 | - | 3 | 506 | 7320690 | ||
FQ224698.1 | - | 3 | 577 |