Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053647.1 | NP_446099.1 | 4 | 588 | ||
GenBank mRNA | AB060092.1 | - | 4 | 588 | ||
U45965.1 | - | 4 | 551 |