Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053656.2 | NP_446108.2 | 2 | 265 | ||
XM_006249515.2 | XP_006249577.1 | 2 | 458 | |||
XM_006249516.2 | XP_006249578.1 | 2 | 457 | |||
XM_006249517.2 | XP_006249579.1 | 2 | 420 | |||
XM_008769305.1 | XP_008767527.1 | 2 | 422 | |||
XM_008769306.1 | XP_008767528.1 | 2 | 134 | |||
XM_008769307.1 | XP_008767529.1 | 2 | 354 | |||
GenBank mRNA | AF020756.1 | - | 2 | 246 | ||
AF020757.1 | - | 2 | 292 | |||
AF020758.1 | - | 1 | 386 | |||
AF020759.1 | - | 2 | 227 | |||
AF064549.1 | - | 2 | 208 | |||
U14414.1 | - | 2 | 265 | |||
Y10473.1 | - | 2 | 265 | |||
Y10474.1 | - | 2 | 247 | |||
Y10475.1 | - | 2 | 301 |