Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053734.2 | NP_446186.2 | 5 | 427 | ||
GenBank mRNA | AF260779.1 | - | 4 | 355 | ||
AF547392.1 | - | 5 | 427 | |||
AF547393.1 | - | 5 | 427 | |||
AY029167.1 | - | 5 | 427 | |||
BC061810.1 | - | 5 | 427 | 5598653 |