Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053785.1 | NP_446237.1 | 5 | 1122 | ||
GenBank mRNA | AF205717.1 | - | 5 | 1122 | ||
BC091131.1 | - | 5 | 937 | 7324973 |