Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037315.1 | NP_001032392.1 | 3 | 471 | ||
NM_001037316.1 | NP_001032393.1 | 2 | 229 | |||
NM_053853.2 | NP_446305.1 | 1 | 624 | |||
XM_006253001.1 | XP_006253063.1 | 3 | 295 | |||
XM_006253002.2 | XP_006253064.1 | 1 | 154 | |||
XM_006253003.1 | XP_006253065.1 | 3 | 299 | |||
XM_006253004.2 | XP_006253066.1 | 3 | 510 | |||
GenBank mRNA | BC078765.1 | - | 2 | 230 | 7127165 | |
U01343.1 | - | 3 | 471 | |||
U01344.1 | - | 1 | 1122 | |||
U17260.1 | - | 2 | 291 |