Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053954.1 | NP_446406.1 | 4 | 1342 | ||
XM_006235509.2 | XP_006235571.1 | 3 | 1758 | |||
GenBank mRNA | Y17606.1 | - | 4 | 1342 |