Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053980.1 | NP_446432.1 | 5 | 460 | ||
XM_006235718.2 | XP_006235780.1 | 5 | 456 | |||
XM_006235720.2 | XP_006235782.1 | 5 | 413 | |||
GenBank mRNA | X78603.1 | - | 5 | 460 |