Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053986.1 | NP_446438.1 | 1487 | |||
GenBank mRNA | X68199.1 | - | 1487 |