Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1PR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005226.3 | NP_005217.2 | 2 | 1279 | ||
GenBank mRNA | AF022139.1 | - | 1 | 845 | ||
AK312798.1 | - | 2 | 1243 | |||
AL832194.1 | - | 1 | 4633 | |||
BC060827.1 | - | 2 | 1241 | 30342032 | ||
BC069579.1 | - | 1 | 847 | 7262431 |
Set Membership: |
G-protein coupled receptor |