Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057131.1 | NP_476472.1 | 3 | 322 | ||
XM_006246434.2 | XP_006246496.1 | 3 | 323 | |||
XM_006246435.2 | XP_006246497.1 | 4 | 529 | |||
XM_006246436.2 | XP_006246498.1 | 3 | 314 | |||
XM_006246437.2 | XP_006246499.1 | 2 | 276 | |||
XM_006246438.2 | XP_006246500.1 | 2 | 197 | |||
XM_006246439.2 | XP_006246501.1 | 3 | 404 | |||
XM_006246440.2 | XP_006246502.1 | 3 | 397 | |||
XM_008767764.1 | XP_008765986.1 | 3 | 395 | |||
XM_008767765.1 | XP_008765987.1 | 2 | 277 | |||
GenBank mRNA | BC078755.1 | - | 1 | 110 | 7113134 | |
D84434.1 | - | 3 | 322 | |||
FQ212452.1 | - | 3 | 270 |