Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057141.1 | NP_476482.1 | 15 | 1552 | ||
XM_006253552.2 | XP_006253614.1 | 17 | 2019 | |||
XM_006253553.2 | XP_006253615.1 | 16 | 1715 | |||
XM_006253554.2 | XP_006253616.1 | 17 | 1959 | |||
XM_006253555.2 | XP_006253617.1 | 16 | 1947 | |||
GenBank mRNA | BC061867.1 | - | 16 | 1699 | 5598872 | |
D17711.1 | - | 15 | 1552 |