Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057197.2 | NP_476545.2 | 2 | 130 | ||
GenBank mRNA | BC059120.1 | - | 2 | 118 | 6920489 | |
CO385941.1 | - | 2 | 115 | |||
D00569.1 | - | 2 | 103 | |||
FQ209689.1 | - | 2 | 119 | |||
FQ210607.1 | - | 2 | 111 | |||
FQ210721.1 | - | 2 | 113 | |||
FQ210830.1 | - | 2 | 100 | |||
FQ212507.1 | - | 2 | 108 | |||
FQ218387.1 | - | 2 | 108 | |||
FQ219629.1 | - | 2 | 111 | |||
FQ219666.1 | - | 2 | 108 | |||
FQ220227.1 | - | 2 | 111 |