Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078623.2 | NP_511178.1 | 8 | 1233 | ||
GenBank mRNA | BC064655.1 | - | 8 | 1233 | 5599860 | |
FQ218103.1 | - | 2 | 463 | |||
FQ218439.1 | - | 7 | 1093 | |||
X15958.1 | - | 8 | 1226 |
Gene Symbol: LOC100911186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748886.2 | XP_003748934.1 | 8 | 1261 |